missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530250
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (86%), Rat (86%), Pig (92%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Phosphoinositide-specific phospholipase C (PLC) acts as a signal transducer that generates 2 second messengers, diacylglycerol and inositol 1, 4, 5-trisphosphate, by hydrolyzing inositol phospholipids. PLC comprises a diverse family of enzymes that differ in structure and tissue distribution (Berridge, 1993 ).
Tekniske data
| PLCD1 | |
| Polyclonal | |
| Unconjugated | |
| PLCD1 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1; AW212592; C79986; NDNC3; phosphoinositide phospholipase C; phosphoinositide phospholipase C-delta-1; phospholipase C delta 1; phospholipase C delta 1 Lf; phospholipase C delta 1 long form; phospholipase C, delta 1; Phospholipase C-delta1; phospholipase C-delta-1; phospholipase C-III; Plc1; Plcd; PLCD1; PLC-delta 1; PLC-delta-1; PLC-III | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5333 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.34 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P51178 | |
| PLCD1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human PLCD1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion