missing translation for 'onlineSavingsMsg'
Learn More

PLC-γ2 (pY759) Mouse, PE, Clone: K86-689.37, BD

Product Code. 15819438
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15819438 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819438 Supplier BD Biosciences Supplier No. 558490

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The Phospholipase C (PLC) isozymes hydrolyze phosphatidyl inositol biphosphate to inositol triphosphate and diacylglycerol. The former causes release of calcium from the endoplasmic reticulum, while the latter is an activator of Protein Kinase C. Within the PLC family, PLC-γ is the only member that contains SH2 and SH3 domains. These domains enable it to interact with protein tyrosine kinases and become enzymatically activated via phosphorylation. It exists as two isoforms: PLC-γ1, which is ubiquitously expressed, and PLC-γ2, which is found primarily in the hematopoietic system. PLC-γ2-null mice exhibit impaired B lymphocyte maturation and defects in Fc receptor functions. Phosphorylation of PLC-γ2 at tyrosines 753 and 759 (Y759) is required for activation of PLC-γ2 enzyme activity. PLC-γ2 phosphorylation at Y759 can be induced by stimulation of the B cell receptor in Ramos cell, the collagen receptor in platelets, and the T cell receptor in Jurkat cells, and it occurs downstream of Btk and BLNK in the signaling cascade of activated B cells. The K86-689.37.73 antibody recognizes PLC-γ2 phosphorylated at Y759 in the SH2-SH3 linker region.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen PLC-γ2 (pY759)
Applications Flow Cytometry
Classification Monoclonal
Clone K86-689.37
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human PLC-γ2 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.