missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKN1, Human, IP-WB Antibody Pair (PW1), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marca: Abnova H00005585-PW1.1Set
Includes: Antibody pair for IP: rabbit polyclonal anti-PKN1 (300μL), Antibody pair for WB: mouse purified polyclonal anti-PKN1 (50μg)
Especificaciones
| PKN1 | |
| Monoclonal/Polyclonal | |
| This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
| DBK/MGC46204/PAK1/PKN/PKN-ALPHA/PRK1/PRKCL1 | |
| Mouse, Rabbit | |
| RUO | |
| 5585 | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PKN1 | |
| PKN1 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-PKN1 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-PKN1 (50 ug) | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido