missing translation for 'onlineSavingsMsg'
Learn More

PKCθ (pT538) Mouse anti-Human, Unlabeled, Clone: 19, BD

Product Code. 15806279
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15806279 50 μg Each
15816279 150 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15806279 Supplier BD Biosciences Supplier No. 612734

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The Protein Kinase C (PKC) family of homologous serine/threonine protein kinases is involved in a number of processes such as growth, differentiation, and cytokine secretion. Three categories exist, conventional PKC (cPKC), novel PKC (nPKC), and atypical PKC (aPKC). These proteins are products of multiple genes and alternative splicing and have different modes of activation. For example, cPKC's members (α, βI, βII, and γ) are calcium activated, phospholipid-dependent serine/threonine specific enzymes which can also be activated by phorbol esters. However, the novel PKC (nPKC) subfamily members (δ , ε, π, and θ isoforms) and the atypical PKC (PKC) subfamily members (ζ , f , andλ isoforms) are Ca2+; independent. The aPKC members are unique in that their activity is independent of diacylglycerols and phorbol esters. The PKC pathway represents a major signal transduction system that is activated following ligand-stimulation of transmembrane receptors by hormones, neurotransmitters and growth factors. PKCθ transcripts are expressed in most tissues with the highest levels being found in hematopoietic tissues and cell lines, including T cells and thymocytes. PKCθ RNA is readily detectable in skeletal muscle, lung, and brain. However, PKCθ expression is not detected in several human carcinoma cell lines. Abundant expression of this PKC isozyme in hematopoietic cells suggests that it may have a role in growth and differentiation processes of these cells. The 9/PKC monoclonal antibody recognizes the phosphorylated threonine 538 (pT538) of human PKCθ.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PKCθ (pT538)
Applications Western Blot
Classification Monoclonal
Clone 19
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human PKCθ Peptide
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.