missing translation for 'onlineSavingsMsg'
Learn More

PKCλ Mouse, Unlabeled, Clone: 41, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610208

 View more versions of this product

Product Code. 15805339

  • ONLINE EXCLUSIVE PRICE
    £412.00
    £396.51 / 1 set
    Save £15.49
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Protein Kinase C (PKC) family of homologous serine/threonine protein kinases is involved in a number of processes such as growth, differentiation, and cytokine secretion. At least eleven isozymes have been described. These proteins are products of multiple genes and alternative splicing. PKC consists of a single polypeptide chain containing four conserved regions (C) and five variable regions (V). The N-terminal half containing C1, C2, V1, and V2 constitutes the regulatory domain and interacts with the PKC activators Ca2+, phospholipid, diacylglycerol, or phorbol ester. However, the novel PKC (nPKC) subfamily members ( δ, ε, η, and θ isoforms) and the atypical PKC (aPKC) subfamily members (ζ, ι, and λ isoforms) are Ca2+ independent and lack the C2 domain. The aPKC members are unique in that their activity is independent of diacylglycerols and phorbol esters. They also lack one repeat of the cysteine-rich sequences that are conserved in cPKC and nPKC. The C-terminal region of PKC contains the catalytic domain. The PKC pathway represents a major signal transduction system that is activated following ligand-stimulation of transmembrane receptors by hormones, neurotransmitters, and growth factors. PKC? shows the highest degree of amino acid homology with PKCζ (72%) and PKCλ mRNA is expressed in a variety of cells and tissues. The PKCλ protein kinase is capable of autophosphorylation and can be activated by phosphatidylserine, but not by other PKC activators such as diacylglycerols, Ca2+, or phorbol esters.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PKCλ
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human PKCγ aa. 397-558
150 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Western Blot
41
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.