Learn More
PKARIIb (pS114) Mouse, Unlabeled, Clone: 47, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 612550
Description
cAMP-dependent Protein Kinase (PKA) is composed of two distinct subunits: catalytic (C) and regulatory (R). Four regulatory subunits have been identified: RIα, RIβ, RIIα dissociate from the catalytic subunits, rendering the enzyme active. Type I and type II holoenzymes have three potential C subunits (Cα, Cβ, or Cγ). Most cells, including T lymphocytes, express both type I and type II PKAs. RIIa expression is associated with cellular transformation, while RIIβ expression correlates with mitotic arrest and cellular differentiation. Type II PKA can be distinguished by autophosphorylation of the R subunits, while type I PKA binds Mg/ATP with high affinity. The cAMP-dependent autophosphorylation of the
human RIIβ subunits occurs at serine 114 (S114). In addition to their enzyme regulatory activity, the RIIα and RIIβ subunits determine the subcellular location of the holoenzymes via their interactions with specific intracellular anchoring proteins. The 47/PKA monoclonal antibody recognizes the phosphorylated S114 in the RIIβ subunit of PKA. The orthologous phosphorylation site in mouse and rat PKA[RIIβ] is S112.
Flow Cytometry, Intracellular Staining, Western Blotting
Specifications
| PKARIIb (pS114) | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Phosphorylated Human PKA[RIIβ] peptide Peptide | |
| 50 μg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 κ |
| Western Blot | |
| 47 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.