missing translation for 'onlineSavingsMsg'
Learn More

PKA (RIIα) Mouse, Unlabeled, Clone: 40, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 612243

 View more versions of this product

Product Code. 15846139

  • ONLINE EXCLUSIVE PRICE
    £297.00
    £284.36 / 1 set
    Save £12.64
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

cAMP-dependent Protein Kinase (PKA) is composed of two distinct subunits: catalytic(C) and regulatory (R). Four regulatory subunits have been identified: RIα, RIβ, RIIα, and RIIβ.These subunits define type I and II cAMP-dependent protein kinases. Following binding of cAMP, the regulatory subunits dissociate from the catalytic subunits, rendering the enzyme active. Type I and type II holoenzymes have three potential C subunits (Cα, Cβ, or Cγ). Type II PKA can be distinguished by autophosphorylation of the R-subunits, while type I PKA binds Mg/ATP with high affinity. Most cells express both type I and type II PKAs. Although the Rα isoforms are ubiquitously expressed, the Rβ isoforms are predominant in nervous and adipose tissues. Expression of the RIα subunit is modulated during muscle and adipocyte differentiation in vitro.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PKA (RIIα)
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human PKA[RIIα] aa. 1-404
150 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Western Blot
40
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.