Learn More
PIR-A/B Rat anti-Mouse, Unlabeled, Clone: 6C1, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 550348
Description
The 6C1 antibody reacts with common epitopes of PIR-A and PIR-B (Paired Immunoglobulin-liked Receptors) in all mouse strains tested (A/J, BALB/cJ, C3H, C57BL/6, DBA/1, DBA/2, NZB, and SJL). PIR-A and PIR-B are type-I transmembrane glycoproteins containing six Ig-like domains. There are multiple PIR-A proteins which are activating receptors by virtue of their intracellular association with the ITAM (Immunoreceptor Tyrosine-based Activation Motif)- containing Fc receptor γ chain (FcRγ) in mast cells and macrophages. FcRγ expression is required for PIR-A cell-surface expression on dendritic cells, B cells, and myeloid lineages. In contrast, PIR-B is an inhibitory receptor which contains various ITIMs (Immunoreceptor Tyrosine-based Inhibitory Motifs) in its cytoplasmic domain. These receptors are expressed on B cells, granulocytes, mast cells, dendritic cells, and monocytes/macrophages, but not on thymocytes, T lymphocytes, erythroid lineage cells, or NK cells. The level of cell-surface expression of these receptors increases as a function of B-cell activation and myeloid- and B-lineage differentiation. The 6C1 antibody immunoprecipitates molecules of 85- and 125kDa, which correspond to PIRA and PIR-B, respectively.
Host Species: Rat
Clone: 6C1
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Recombinant mouse PIR-A1 extracellular domain peptide and the mouse myeloid cell line, WEH13
Flow Cytometry, Immunoprecipitation
Specifications
| PIR-A/B | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant mouse PIR-A1 extracellular domain peptide and the mouse myeloid cell line, WEH13 | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| 6C1 | |
| Unconjugated | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.