missing translation for 'onlineSavingsMsg'
Learn More

PIN1 Polyclonal Antibody, Invitrogen™

Product Code. 11545862
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11545862 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11545862 Supplier Invitrogen Supplier No. PA110013

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chicken Polyclonal Antibody

Peptidyl-prolyl cis/trans isomerases , such as PIN1, catalyze the cis/trans isomerization of peptidyl-prolyl peptide bonds. PIN1 is the only PPIase that specifically binds to phosphorylated ser/thr-pro motifs to catalytically regulate the post-phosphorylation conformation of its substrates. PIN1-catalyzed conformational regulation has a profound impact on key proteins involved in the regulation of cell growth, genotoxic and other stress responses, the immune response, germ cell development, neuronal differentiation, and survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIN1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 10mM sodium azide
Gene PIN1
Gene Accession No. Q5BIN5, Q13526, Q9QUR7
Gene Alias 0610025L01Rik, D9Bwg1161e, Peptidyl-prolyl cis/trans isomerase
Gene Symbols PIN1
Host Species Chicken
Immunogen Purified, recombit human Pin-1.
Purification Method PEG precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100055392, 100512827, 100859153, 23988, 298696, 5300, 535470
Target Species Bovine, Chicken, Equine, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.