missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PILRA Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547613
This item is not returnable.
View return policy
Description
In Western blots, approximately 20% cross-reactivity with recombinant mouse PILR-beta is observed and 5% cross-reactivity with recombinant human PILR-alpha is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
PILRA Polyclonal antibody specifically detects PILRA in Mouse samples. It is validated for Flow Cytometry, Western Blot.Specifications
PILRA | |
Polyclonal | |
Unconjugated | |
PILRA | |
AV021745; cell surface receptor FDF03; FDF03; Inhibitory receptor PILR-alpha; paired immunoglobin like type 2 receptor alpha; paired immunoglobin-like type 2 receptor alpha; paired immunoglobulin-like type 2 receptor alpha; Pilra | |
Goat | |
Antigen affinity chromatography | |
RUO | |
231805 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q2YFS3 | |
PILRA | |
Mouse myeloma cell line NS0-derived recombinant mouse PILR-alpha isoform 1 Leu21-Val197 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |