missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-PIGH, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00005283-PW1.1Set
Additional Details : Weight : 0.02000kg
Includes: Antibody pair for IP: rabbit polyclonal anti-PIGH (300μL), Antibody pair for WB: mouse purified polyclonal anti-PIGH (50μg)
Specifications
PIGH | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
GPI-H | |
Mouse, Rabbit | |
RUO | |
5283 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
PIGH | |
PIGH | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-PIGH (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-PIGH (50 ug) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only