Learn More
PIAS1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PIAS1 protein.
Brand: Abnova H00008554-B01.50uL
Description
This gene encodes a member of the mammalian PIAS [protein inhibitor of activated STAT-1 (signal transducer and activator of transcription-1)] family. This member contains a putative zinc-binding motif and a highly acidic region. It inhibits STAT1-mediated gene activation and the DNA binding activity, binds to Gu protein/RNA helicase II/DEAD box polypeptide 21, and interacts with androgen receptor (AR). It functions in testis as a nuclear receptor transcriptional coregulator and may have a role in AR initiation and maintenance of spermatogenesis. [provided by RefSeq
Sequence: MADSAELKQMVMSLRVSELQVLLGYAGRNKHGRKHELLTKALHLLKAGCSPAVQMKIKELYRRRFPQKIMTPADLSIPNVHSSPMPATLSPSTIPQLTYDGHPASSPLLPVSLLGPKHELELPHLTSALHPVHPDIKLQKLPFYDLLDELIKPTSLASDNSQRFRETCFAFALTPQQVQQISSSMDISGTKCDFTVQVQLRFCLSETSCPQEDHFPPNLCVKVNTKPCSLPGYLPPTKNGVEPKRPSRPINITSLVRLSTTVPNTIVVSWTAEIGRNYSMAVYLVKQLSSTVLLQRLRAKGIRNPDHSRALIKEKLTADPDSEIATTSLRVSLLCPLGKMRLTIPCRALTCSHLQCFDATLYIQMNEKKPTWVCPVCDKKAPYEHLIIDGLFMEILKYCTDCDEIQFKEDGTWAPMRSKKEVQEVSASYNGVDGCLSSTLEHQVASHHQSSNKNKKVEVIDLTIDSSSDEEEEEPSAKRTCPSLSPTSPLNNKGILSLPHQASPVSRTPSLPAVDTSYINTSLIQDYRHPFHMTPMPYDLQGLDFFPFLSGDNQHYNTSLLAAAAAAVSDDQDLLHSSRFFPYTSSQMFLDQLSAGGSTSLPTTNGSSSGSNSSLVSSNSLRESHSHTVTNRSSTDTASIFGIIPDIISLDSpecifications
| PIAS1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human PIAS1 protein. | |
| PIAS1 | |
| DDXBP1/GBP/GU/RH-II/MGC141878/MGC141879/ZMIZ3 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 8554 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_016166.1 | |
| PIAS1 | |
| PIAS1 (NP_057250.1, 1 a.a. ∼ 651 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.