Learn More
PI3-Kinase p170 Mouse, Unlabeled, Clone: 17, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 611046
Description
PI3-kinase phosphorylates the D-3 position of the inositol ring of phosphatidylinositol (PtdIns), producing PtdIns(3)P, PtdIns(3,4)P2, and PtdIns(3,4,5)P3. PI3-Kinase is a heterodimer of an 85kDa regulatory subunit (p85) and a 110kDa catalytic subunit (p110). However, this heterodimer is only one member of a larger family of proteins with similarity to the p110 subunit. These different PI3-kinase isoforms have been divided into three classes. Class I consists of p110α and p110β which bind the p85 subunit and associate with receptor tyrosine kinases. Class II contains 68D, cpk, and p170. These proteins all contain a C-terminal C2 domain and phosphorylate PtdIns and PtdIns(4)P, but not
PtdIns(4,5)P2. Class III members only phosphorylate PtdIns to PtdIns(3)P and include the S. cerevisiae Vps34p and its human homologs. The first human Class II PI3-kinase to be described, p170, contains the characteristic C-terminal phosphoinositide-binding C2 domain also found in PKC isoforms and synaptotagmins. In addition, expression of PI3-kinase p170 is widespread and may indicate that the protein performs a housekeeping function in cell growth and differentiation.
Immunofluorescence, Western Blotting
Specifications
| PI3-Kinase p170 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Mouse PI3-Kinase p170 | |
| 50 μg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 17 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.