missing translation for 'onlineSavingsMsg'
Learn More

PI3-Kinase p110α Mouse, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

£237.00 - £284.36

Specifications

Antigen PI3-Kinase p110α
Clone 19
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15855919
missing translation for 'viewDocuments'
BD Biosciences
611398
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
15865919
missing translation for 'viewDocuments'
BD Biosciences
611399
150 μg
ONLINE EXCLUSIVE PRICE
£297.00
£284.36 / 1 set
Save £12.64
4% Off
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phosphatidylinositol 3 (PI3) -kinase participates in insulin-stimulated glucose uptake, PDGF-induced membrane ruffling, and G-protein receptor signaling. It exists as a heterodimer of 85kDa (p85) and 110kDa (p110) subunits. The p85 subunit associates with and serves as a substrate for activated growth factor receptor tyrosine kinases. p85 regulates the p110 catalytic subunit by acting as the link between PI3-kinase and the ligand-activated receptor. Four isoforms of p110 have been identified (α, β, γ, and δ). The p110α isoform contains an N-terminal region involved in p85 binding and a C-terminal kinase domain. p85/p110α-type PI kinase phosphorylates the D-3 and D-4
position of the inositol ring of PI, thereby producing PtdIns(3)P, PtdIns(3,4)P[2], PtdIns(3,4,5)P[3], PtdIns(4)P, and PtdIns(4,5) P[2]. During induction of chemotaxis by the chemokine SDF-1α, PI3-kinase regulates adhesion and ERM protein redistribution in the lymphocyte plasma membrane. In addition, PI3-kinases activate other signaling molecules, such as p70 S6 kinase and Akt/protein kinase B. Thus, p85/p110α-type PI kinase is a ubiquitously expressed kinase that is involved in a variety of cell signaling cascades.

Host Species: Mouse
Clone: 19
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human PI3-Kinase p110α aa. 101-300
Formula Weight [Chemical]: 110kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

PI3-Kinase p110α
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
19
Western Blot
Unconjugated
RUO
Human, Rat
Human PI3-Kinase p110α aa. 101-300
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.