Learn More
anti-Phosphotyrosine, Polyclonal, Abcam™
Brand: Abcam AB9319-100UG
Description
Host Species: Rabbit
Isotype: IgG
Immunogen: Chemical / Small Molecule conjugated to KLH.
Applications:
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
| Phosphotyrosine | |
| .25mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Neuroscience, Signal Transduction | |
| IgG |
| Polyclonal | |
| Unconjugated | |
| Chemical / Small Molecule conjugated to KLH. | |
| 100 μg | |
| Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.