Learn More
Phosphothreonine/pT Polyclonal Antibody, Invitrogen™
Phosphothreonine/pT Antibody, 71-8200, from Invitrogen™. Species Reactivity: Chemical; Applications: ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Brand: Invitrogen 718200
Description
Phosphothreonine/pT Antibody, 71-8200, from Invitrogen™. Species Reactivity: Chemical; Applications: ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Host Species: Rabbit
Cross Reactivity: Chemical
Isotype: IgG
Immunogen: Phosphothreonine containing proteins.
Formulation: 50μg purified antibody (0.25mg/mL) in PBS and 0.1% sodium azide; pH 7.4
Synonyms: Threonine, pThr
References:
Test: ELISA, Species: Hu, Reference: High-throughput nonradioisotopic detection of picomole levels of phosphothreonine and phosphoserine containing peptides via biotinylation and enzyme-linked immunosorbent assay. (Mar 1999) Anal. Biochem., Vol: 268, 2, 371-6, Mahoney CW,Hosoi T,Ohashi M; Pubmed ID: 10075828
Specifications
| Phosphothreonine/pT | |
| Polyclonal | |
| Threonine, pThr | |
| Phosphothreonine containing proteins. | |
| 50 μg | |
| Store at -20°C | |
| IgG |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity chromatography | |
| Primary | |
| Purified | |
| Chemical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.