missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522323
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, HeLa, HepG2. Predicted reactivity: Mouse (93%), Rat (95%), Chicken (86%), Rhesus Monkey (98%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene catalyzes the isomerization of mannose 6-phosphate to mannose 1-phosphate, which is a precursor to GDP-mannose necessary for the synthesis of dolichol-P-oligosaccharides. Mutations in this gene have been shown to cause defects in glycoprotein biosynthesis, which manifests as carbohydrate-deficient glycoprotein syndrome type I.
Specifications
| PMM2 | |
| Polyclonal | |
| Unconjugated | |
| PMM2 | |
| AI585868; C86848; CDG1; CDG1a; CDGS; mannose-6-phosphate isomerase; phosphomannomutase 2; phosphomannose isomerase 1; PMI; PMI1; PMM 2; Pmm2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5373 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O15305 | |
| PMM2 | |
| Recombinant protein encompassing a sequence within the center region of human PMM2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction