missing translation for 'onlineSavingsMsg'
Learn More

Phospho-WASP (Tyr290) Polyclonal Antibody, Invitrogen™

Product Code. 15535271
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15535271 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15535271 Supplier Invitrogen Supplier No. PA538346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Effector protein for Rho-type GTPases. Regulates actin filament reorganization via its interaction with the Arp2/3 complex. Important for efficient actin polymerization. Possible regulator of lymphocyte and platelet function. Mediates actin filament reorganization and the formation of actin pedestals upon infection by pathogenic bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-WASP (Tyr290)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WAS
Gene Accession No. P42768, P70315
Gene Alias IMD2, SCNX, THC, THC1, Thrombocytopenia 1 (X linked), U42471, Was, WASp, WASP, Wiskott Aldrich syndrome (eczema thrombocytopenia), Wiskott Aldrichsyndrome protein, Wiskott-Aldrich syndrome protein
Gene Symbols WAS
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human WASP around the phosphorylation site of Tyr290 (L-I-YP-D-F)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22376, 7454
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.