missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-VDR (Ser208) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538931
This item is not returnable.
View return policy
Description
Nuclear hormone receptor. Transcription factor that mediates the action of vitamin D3 by controlling the expression of hormone sensitive genes. Regulates transcription of hormone sensitive genes via its association with the WINAC complex, a chromatin-remodeling complex. Recruited to promoters via its interaction with the WINAC complex subunit BAZ1B/WSTF, which mediates the interaction with acetylated histones, an essential step for VDR-promoter association. Plays a central role in calcium homeostasis.
Specifications
| Phospho-VDR (Ser208) | |
| Polyclonal | |
| Unconjugated | |
| VDR | |
| 1,25-dihydroxyvitamin D3 receptor, NR1I1, vitamin D receptor, vitamin D3 receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7421 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11473 | |
| VDR | |
| A synthetic phosphopeptide derived from human Vitamin D Receptor around the phosphorylation site of Ser208 (D-L-SP-E-E) | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction