missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PLCG2 (Tyr1217) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538319
This item is not returnable.
View return policy
Description
Phosphoinositide-specific phospholipase C (PLC) plays a crucial role in the initiation of receptor mediated signal transduction through the generation of the two second messengers, inositol 1,4,5-triphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. There are many mammalian PLC isozymes, including PLC beta1, PLC beta2, PLC beta3, PLC beta4, PLC gamma1, PLC gamma2, PLC delta1, PLC delta2 and PLCe. PLC delta exists as four different isoforms. PLC delta1, a calcium signal amplifier, is activated by an atypical GTP-binding protein. In addition, PLC delta1 is an effector for GTP-binding protein transglutaminase II-mediated oxytocin receptor and alpha1B-adrenoreceptor signaling. Mouse PLC delta1 is highly expressed in brain, heart, lung and testis. PLC delta is abnormally accumulated in autopsied brains with Alzheimer′s disease (AD), suggesting that it may play a role in the pathology of AD. PLC delta2 is markedly expressed in type II intestinal metaplasia and in the adenocarcinoma. When PLC delta2 is expressed in type I intestinal metaplasia, the metaplasia is generally considered benigt, yet evolves toward neoplastic transformation. Thus, PLC delta2 expression may be a possible marker of gastric maligt transformation.
Specifications
| Phospho-PLCG2 (Tyr1217) | |
| Polyclonal | |
| Unconjugated | |
| PLCG2 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-2, Phosphoinositide phospholipase C-gamma-2, Phospholipase C-IV, PLC-IV, Phospholipase C-gamma-2, PLC-gamma-2, PLCG2, PLC gamma2, PLC- gamma2, PLC gamma-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 234779, 29337, 5336 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P16885, Q8CIH5, P24135 | |
| PLCG2 | |
| A synthetic phosphopeptide derived from human PLCG2 around the phosphorylation site of Tyr1217 (L-F-L-YP-D-T) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction