missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Fyn (Tyr530) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538115
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Fyn is a membrane-associated, non-receptor protein tyrosine kinase approximately 59kDa, which belongs to the Src family of cytoplasmic tyrosine kinases. Fyn is very strongly similar to mouse Fyn, v-yes and c-src. Fyn is expressed predominately in tissues of neuronal and hematopoietic origin. Neuronal Fyn and hematopoietic Fyn differ at the junction of the SH2 and kinase domains due to tissue specific alternative splicing. Fyn has been shown to be involved in B cell and T cell activation as well as keratinocyte differentiation. In T cells, Fyn associates with the T-cell antigen receptor and Thy1.Specifications
Phospho-Fyn (Tyr530) | |
Polyclonal | |
Unconjugated | |
FYN | |
EC 2.7.10.2, FYN, P59-FYN, SLK, SYN, kinase Fyn | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14360, 2534 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P06241, P39688 | |
FYN | |
A synthetic phosphopeptide derived from human Fyn around the phosphorylation site of Tyr530 (P-Q-YP-Q-P) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |