missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-NFAT5 (Ser155) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539799
This item is not returnable.
View return policy
Description
The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
Specifications
| Phospho-NFAT5 (Ser155) | |
| Polyclonal | |
| Unconjugated | |
| NFAT5 | |
| Nuclear Factor Activated T-Cells, NFATL1, Nuclear factor of activated T cells 5, T cell transcription factor NFAT5, NF-AT5, Tonicity-responsive enhanc | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10725, 54446 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O94916, Q9WV30 | |
| NFAT5 | |
| A synthetic phosphopeptide derived from human NFAT5/TonEBP around the phosphorylation site of Ser155 (D-N-SP-R-M) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction