missing translation for 'onlineSavingsMsg'
Learn More

Phospho-MUNC18 (Ser313) Polyclonal Antibody, Invitrogen™

Product Code. 15535221
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15535221 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15535221 Supplier Invitrogen Supplier No. PA538296

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-MUNC18 (Ser313)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene STXBP1
Gene Accession No. P61764, O08599, P61765
Gene Alias STXBP1, MUNC18-1, UNC18, hUNC18, syntaxin binding protein 1, Hunc18, rbSec1, syntaxin-binding protein 1, RP11-56D16.3, EIEE4, FLJ37475, RBSEC1
Gene Symbols STXBP1
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human MUNC-18a around the phosphorylation site of Ser313 (S-S-SP-K-R)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20910, 25558, 6812
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.