missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MCM4 (Ser54) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536859
Code : NEW
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 90 kDa. Purity is >95% by SDS-PAGE.
Phospho-MCM4 (Ser54) Polyclonal antibody specifically detects Phospho-MCM4 (Ser54) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-MCM4 (Ser54) | |
Polyclonal | |
Unconjugated | |
Mcm4 | |
19G; AI325074; AU045576; cb1025; Cdc21; CDC21 homolog; CDC54; DNA replication licensing factor MCM4; fc12c09; fj85g09; hCdc21; hm:zeh1616; homolog of S. pombe cell devision cycle 21; mcdc21; MCM4; MCM4 minichromosome maintenance deficient 4; MCM4 minichromosome maintenance deficient 4, mitotin; mcm4 protein; Mcmd4; mini chromosome maintenance deficient 4 homolog; minichromosome maintenance complex component 4; minichromosome maintenance deficient 4; minichromosome maintenance deficient 4 homolog; minichromosome maintenance deficient 4 homolog (S. cerevisiae); mKIAA4003; NKCD; NKGCD; P1-CDC21; Unknown (protein for MGC:140484); wu:fc12c09; wu:fj85g09; zeh1616 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17217, 29728, 4173 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P33991, P49717 | |
Mcm4 | |
Synthetic phosphopeptide derived from human MCM4 around the phosphorylation site of Serine 54 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |