missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-HDAC4/HDAC5/HDAC9 (Ser246, Ser259, Ser220) Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA537835

Product Code. 15564771

  • £442.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A suggested positive control for Western blot is 293 cells; suggested positive control for IHC is human breast carcinoma.

Histone deacetylase (HDAC) and histone acetyltransferase (HAT) are enzymes that regulate transcription by selectively deacetylating or acetylating the eta-amino groups of lysines located near the amino termini of core histone proteins (1). Eight members of HDAC family have been identified in the past several years (2,3). These HDAC family members are divided into two classes, I and II. Class I of the HDAC family comprises four members, HDAC-1, 2, 3, and 8, each of which contains a deacetylase domain exhibiting from 45 to 93% identity in amino acid sequence. Class II of the HDAC family comprises HDAC-4, 5, 6, and 7, the molecular weights of which are all about twofold larger than those of the class I members, and the deacetylase domains are present within the C-terminal regions, except that HDAC-6 contains two copies of the domain, one within each of the N-terminal and C-terminal regions. Human HDAC-1, 2 and 3 were expressed in various tissues, but the others (HDAC-4, 5, 6, and 7) showed tissue-specific expression patterns (3). These results suggested that each member of the HDAC family exhibits a different, individual substrate specificity and function in vivo.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-HDAC4/HDAC5/HDAC9 (Ser246, Ser259, Ser220)
Polyclonal
Unconjugated
HDAC4
AHO3; Antigen NY-CO-9; BDMR; DKFZp779K1053; HA6116; HD4; HD5; HD7; HD7b; HD9; HDAC; HDAC4; HDAC-4; HDAC5; HDAC7; HDAC7B; HDAC9; hdac-9; HDAC9B; HDAC9FL; HDACA; HDAC-A; HDRP; histone deacetylase 4; histone deacetylase 4/5-related protein; histone deacetylase 5; Histone deacetylase 7B; histone deacetylase 9; histone deacetylase A; Histone deacetylase-related protein; KIAA0288; KIAA0600; KIAA0744; MEF-2 interacting transcription repressor (MITR) protein; MEF2-interacting transcription repressor MITR; MITR; NY-CO-9
Rabbit
Antigen affinity chromatography
RUO
10014, 9734, 9759
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P56524, Q9UKV0, Q9UQL6
HDAC4, HDAC5, HDAC9
Peptide sequence around phosphorylation site of serine 246/259/220 (T-A-S(p)-EP) derived from Human HDAC4/HDAC5/HDAC9.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.