Learn More
anti-Phospho-GSK3 beta pSer9 Oligoclonal, Invitrogen™
Phospho-GSK3 beta pSer9Oligoclonal antibody 710100 from Thermo Scientific™ Pierce™ specifically detects Phospho-GSK3 beta pSer9 in Human samples, and it is validated for WB and ChIP
Brand: Invitrogen 710100
Description
Glycogen synthase kinase-3 beta (GSK3 beta) belongs to the subfamily of glycogen synthase kinases, that play a central role in energy metabolism, neuronal cell development, and body pattern formation. GSK3 has alpha and beta isoforms (51 kDa and 47 kDa respectively), that are functionally dissimilar. GSK3 beta plays a role in insulin and Wnt signaling, and is inhibited following the extracellular signaling events.
710100 detects Phospho-GSK3 beta pSer9 in Human samples. 710100 has been successfully used in WB and ChIP procedures. The immunogen for 710100 is phosphopeptide corresponding to amino acids 5-13 aa of human GSK3 beta.
Applications:
Chromatin Immunoprecipitation, Western Blotting
Specifications
| Phospho-GSK3 beta pSer9 | |
| 9HCLC | |
| P49841 | |
| Phosphopeptide corresponding to amino acids 5-13 aa of human GSK3 beta | |
| Primary | |
| Human | |
| Purified |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Protein A | |
| 2932 | |
| Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.