missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-DDX5 (Tyr593) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538471
This item is not returnable.
View return policy
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a RNA-dependent ATPase, and also a proliferation-associated nuclear antigen, specifically reacting with the simian virus 40 tumor antigen. This gene consists of 13 exons, and alternatively spliced transcripts containing several intron sequences have been detected, but no isoforms encoded by these transcripts have been identified.
Specifications
| Phospho-DDX5 (Tyr593) | |
| Polyclonal | |
| Unconjugated | |
| DDX5 | |
| G17P1, HLR1, HUMP68, p68, ATP-dependent RNA helicase DDX5, DEAD box protein 5, DEAD box-5, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 5 (RNA helicase, 68kD), RNA helicase p68, probable ATP-dependent RNA helicase DDX5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13207, 1655 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17844, Q61656 | |
| DDX5 | |
| A synthetic phosphopeptide derived from human DDX5/DEAD-box Protein 5 around the phosphorylation site of Tyr593 (Q-A-YP-A-Y) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction