missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Cdc25A (Ser79) Polyclonal Antibody
GREENER_CHOICE

Product Code. 10803801
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10803801 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10803801 Supplier Invitrogen™ Supplier No. PA512951

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

CDC25a is a member of the CDC25 family of phosphatases. Three CDC25 genes are present in human cells: CDC25A, CDC25B, and CDC25C. These three genes function at different phases of the cell cycle. CDC25a is required for progression through G1 and S phases of the cell cycle. CDC25a activates the cyclin-dependent kinase CDC2 by removing two phosphate groups. CDC25A is specifically degraded in response to DNA damage, which prevents cells with chromosomal abnormalities from progressing through cell division. CDC25A is an oncogene, although its exact role in oncogenesis has not been demonstrated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Cdc25A (Ser79)
Applications Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CDC25A
Gene Accession No. P30304
Gene Alias CDC25 isoform A1-CAG; CDC25A; CDC25A2; CDC25A2-CAG isoform; Cdc25m3; cell division cycle 25A; D9Ertd393e; dual specificity phosphatase Cdc25A; EC 3.1.3.48; M-phase inducer phosphatase 1; MPIP1
Gene Symbols CDC25A
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S79 of human Cdc25A.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 993
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.