missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-CCR5 (Ser349) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538081
This item is not returnable.
View return policy
Description
Receptors for the CC chemokine family include CCR1, CCR2A, CCR3, CCR4, CCR5 and the Duffy blood group antigen. The CC receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation. The HIV receptor is composed of two molecules, the CD4 binding receptor and a co receptor. The chemokine receptor CCR5 is one of the co receptors used by primary isolates of HIV1. Extracellular domains of CCR5 are important for HIV entry into the target cells.
Specifications
| Phospho-CCR5 (Ser349) | |
| Polyclonal | |
| Unconjugated | |
| CCR5 | |
| C-C CKR-5, C-C chemokine receptor type 5, CC-CKR-5, CCR-5, CD195 antigen, CHEMR13, CKR5, CMKBR5, HIV-1 fusion co-receptor, MIP-1 alpha receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1234 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51681 | |
| CCR5 | |
| A synthetic phosphopeptide derived from human CCR5 around the phosphorylation site of Ser349 (E-I-SP-V-G) | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction