missing translation for 'onlineSavingsMsg'
Learn More

Phospho-CaMKIV (Thr196, Thr200) Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Product Code. 15514041
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15514041 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15514041 Supplier Invitrogen Supplier No. PA536745

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous levels of human CaMKIV protein at a molecular weight of 60 kDa only when phosphorylated at Thr200, and detects mouse and rat CaMKIV protein when phosphorylated at the corresponding site Thr196. Purity is >95% by SDS-PAGE.

Ca2+/calmodulin dependent protein kinase type IV (CAMKIV) belongs to the serine/threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This enzyme is a multifunctional serine/threonine protein kinase with limited tissue distribution, that has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-CaMKIV (Thr196, Thr200)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CAMK4
Gene Accession No. Q16566, P08414, P13234
Gene Alias Calcium/calmodulin-dependent protein kinase type IV, CaMK IV, CaM kinase-GR, CAMK4, CAMK, CAMK-GR, CAMKIV
Gene Symbols CAMK4
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from human CaMKIV around the phosphorylation site of Threonine 196/200
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12326, 25050, 814
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.