missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-CaMKIV (Thr196, Thr200) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538435
This item is not returnable.
View return policy
Description
Ca2+/calmodulin dependent protein kinase type IV (CAMKIV) belongs to the serine/threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This enzyme is a multifunctional serine/threonine protein kinase with limited tissue distribution, that has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells.
Specifications
| Phospho-CaMKIV (Thr196, Thr200) | |
| Polyclonal | |
| Unconjugated | |
| CAMK4 | |
| Calcium/calmodulin-dependent protein kinase type IV, CaMK IV, CaM kinase-GR, CAMK4, CAMK, CAMK-GR, CAMKIV | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12326, 25050, 814 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16566, P08414, P13234 | |
| CAMK4 | |
| A synthetic phosphopeptide derived from human CaMK4 around the phosphorylation site of Thr196/200 (M-K-TP-V-C) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction