missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-c-Jun (Ser243) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517261
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
c-Jun is a transcription factor that recognizes and binds to the enhancer heptamer motif 5'-TGA[CG]TCA-3'. It promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation. Involved in activated KRAS-mediated transcriptional activation of USP28 in colorectal cancer (CRC) cells.Specifications
Phospho-c-Jun (Ser243) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P05412, P05627, P17325 | |
JUN | |
Synthetic phosphopeptide corresponding to residues around pSer243 of human c-Jun | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
JUN | |
Activator protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; cjun; c-Jun; c-jun transcription factor; enhancer-binding protein AP1; I79_009953; immediate early; JUN; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; p39; proto-oncogene c-Jun; Rjg-9; transcription factor; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16476, 24516, 3725 | |
-20°C | |
Liquid |