missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-alpha Synuclein (Ser129) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537740
This item is not returnable.
View return policy
Description
A suggested positive control for Western blot is mouse brain cells.
Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. These proteins are abundant in the brain and play a role in regulating synaptic transmission and membrane trafficking. Alpha- and beta-synuclein have been shown to selectively inhibit phospholipase D2, suggesting that they may also be involved in signaling pathways. Defects in the SNCA gene, which encodes alpha-synuclein, have been implicated in the pathogenesis of Parkinson's disease. Additionally, alpha-synuclein peptides are a major component of amyloid plaques in the brains of patients with Alzheimer's disease. Multiple alternatively spliced transcripts encoding different isoforms have been identified for this gene. Diseases associated with SNCA include Parkinson Disease 1, Autosomal Dominant and Dementia, Lewy Body.
Specifications
| Phospho-alpha Synuclein (Ser129) | |
| Polyclonal | |
| Unconjugated | |
| SNCA | |
| Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20617, 29219, 6622 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O55042, P37377, P37840 | |
| SNCA | |
| Peptide sequence around phosphorylation site of serine 129 (M-P-S(p)-E-E) derived from Human alpha-Synuclein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction