missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-ABL1/ABL2 (Tyr393, Tyr439) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536821
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 123 kDa. Purity is >95% by SDS-PAGE.
The Abl oncogene was initially identified as the viral transforming gene of Abelson murine leukemia virus (A-MuLV). The major translational product of c-Abl has been identified as a protein with tyrosine kinase activity and an SH2 domain. The Abl oncogene is implicated in several human leukemias including 90-95% of chronic myelocytic leukemia (CML), 20-25% of adult acute lymphoblastic leukemia (ALL) and 2-5% of pediatric ALL. In these leukemias the c-Abl proto-oncogene undergoes a (9;22) chromosomal translocation producing the Philadelphia (Ph1) chromosome. The molecular consequence of this translocation is the generation of a chimeric Bcr/c-Abl mRNA encoding activated Abl protein-tyrosine kinase.
Specifications
| Phospho-ABL1/ABL2 (Tyr393, Tyr439) | |
| Polyclonal | |
| Unconjugated | |
| ABL1 | |
| AA536808; Abelson murine leukemia oncogene; Abelson murine leukemia viral (v-abl) oncogene homolog 1; Abelson murine leukemia viral (v-abl) oncogene homolog 2; abelson murine leukemia viral oncogene homolog 1; Abelson murine leukemia viral oncogene homolog 2; Abelson tyrosine-protein kinase 1; Abelson tyrosine-protein kinase 2; Abelson-related gene protein; Abl; ABL proto-oncogene 1, non-receptor tyrosine kinase; ABL proto-oncogene 2, non-receptor tyrosine kinase; ABL1; Abl1-related gene product; abl2; ABLL; AI325092; Arg; arg tyrosine kinase; bcr/abl; bcr/c-abl oncogene protein; c-Abl; c-abl oncogene 1, non-receptor tyrosine kinase; c-abl oncogene 1, receptor tyrosine kinase; c-abl oncogene 2, non-receptor tyrosine kinase; c-ABL1; E430008G22Rik; JTK7; p150; Proto-oncogene c-Abl; proto-oncogene tyrosine-protein kinase ABL1; tyrosine kinase ARG; tyrosine-protein kinase ABL1; Tyrosine-protein kinase ABL2; tyrosine-protein kinase ARG; v-abl; v-abl Abelson murine leukemia oncogene 1; v-abl Abelson murine leukemia viral oncogene 1; v-abl Abelson murine leukemia viral oncogene 2; v-abl Abelson murine leukemia viral oncogene 2 (arg, Abelson-related gene); v-abl Abelson murine leukemia viral oncogene homolog 1; v-abl Abelson murine leukemia viral oncogene homolog 2; v-abl Abelson murine leukemia viral oncogene homolog 2 (arg, Abelson-related gene) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11350, 11352, 25, 27, 304883, 311860 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P00519, P00520, P42684, Q4JIM5 | |
| ABL1, ABL2 | |
| Synthetic phosphopeptide derived from human ABL1/2 around the phosphorylation site of Tyrosine 393/439. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction