Learn More
pleckstrin homology-like domain, family A, member 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human PHLDA1 protein.
Brand: Abnova H00022822-D01P.100ug
Description
This gene encodes an evolutionarily conserved proline-histidine rich nuclear protein. The encoded protein may play an important role in the anti-apoptotic effects of insulin-like growth factor-1. [provided by RefSeq
Sequence: MLESSGCKALKEGVLEKRSDGLLQLWKKKCCILTEEGLLLIPPKQLQHQQQQQQQQQQQQQQPGQGPAEPSQPSGPAVASLEPPVKLKELHFSNMKTVDCVERKGKYMYFTVVMAEGKEIDFRCPQDQGWNAEITLQMVQYKNRQAILAVKSTRQKQQHLVQQQPPSQPQPQPQLQPQPQPQPQPQPQPQSQPQPQPQPKPQPQQLHPYPHPHPHPHSHPHSHPHPHPHPHPHQIPHPHPQPHSQPHGHRLLRSTSNSASpecifications
| pleckstrin homology-like domain, family A, member 1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human PHLDA1 protein. | |
| PHLDA1 | |
| DT1P1B11/MGC131738/PHRIP/TDAG51 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC018929.2 | |
| PHLDA1 | |
| PHLDA1 (AAH18929.3, 1 a.a. ∼ 259 a.a) full-length human protein. | |
| 100 μg | |
| Apoptosis | |
| 22822 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.