Learn More
anti-PGC1α, Polyclonal, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Brand: Abcam AB54481-50UL
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Human, Zebrafish. Predicted to work with Rat, Horse, Chicken, Cow, Dog, Pig, Cynomolgus Monkey, Rhesus monkey, Opossum
Immunogen: Synthetic peptide: C-SKYDSLDFDSLLKEAQRSLRR, corresponding to amino acids 777-797 of Human PGC1 alpha
Applications:
ELISA, Immunocytochemistry, Western Blotting
Specifications
| PGC1α | |
| 1mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Cardiovascular, Epigenetics And Nuclear Signaling | |
| Bovine, Canine, Chicken, Cynomolgus, Equine, Human, Monkey, Mouse, Opossum, Pig, Rat, Rhesus Monkey, Zebrafish |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide: C-SKYDSLDFDSLLKEAQRSLRR, corresponding to amino acids 777-797 of Human PGC1 alpha | |
| 50 μL | |
| Primary | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.