missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGC1 alpha Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538022
This item is not returnable.
View return policy
Description
A recommended positive control is human heart tissue lysate. At least three isoforms of PPARGC1A are known to exist; this antibody will detect all three isoforms.
PGC1 alpha is a transcriptional coactivator that regulates the genes involved in energy metabolism. It interacts with PPARgamma, which permits it′s interaction with multiple transcription factors. PGC-1 Alpha can interact with, and regulate the activities of, cAMP response element binding protein (CREB) and nuclear respiratory factors (NRFs). It provides a direct link between external physiological stimuli and the regulation of mitochondrial biogenesis, and is a major factor that regulates muscle fiber type determination. It may be also involved in controlling blood pressure, regulating cellular cholesterol homoeostasis, and the development of obesity.
Specifications
| PGC1 alpha | |
| Polyclonal | |
| Unconjugated | |
| PPARGC1A | |
| PPARGC1A, LEM6, PGC1, PGC1A, PGC-1v, PPARGC1, LEM6, Peroxisome proliferator-activated receptor gamma coactivator 1-alpha, Ligand effect modulator 6, PGC-1-alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10891, 19017, 83516 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9UBK2, O70343, Q9QYK2 | |
| PPARGC1A | |
| 17 amino acid peptide near the amino terminus of human PPARGC1A | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction