Learn More
PGC1 alpha Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA538021
Product Code. 15554951
  • £441.00 / 100µg
Description
A recommended positive control is mouse liver tissue lysate. At least three isoforms of PPARGC1A are known to exist; this antibody only detects the longest isoform.
PGC1 alpha is a transcriptional coactivator that regulates the genes involved in energy metabolism. It interacts with PPARgamma, which permits it′s interaction with multiple transcription factors. PGC-1 Alpha can interact with, and regulate the activities of, cAMP response element binding protein (CREB) and nuclear respiratory factors (NRFs). It provides a direct link between external physiological stimuli and the regulation of mitochondrial biogenesis, and is a major factor that regulates muscle fiber type determination. It may be also involved in controlling blood pressure, regulating cellular cholesterol homoeostasis, and the development of obesity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PGC1 alpha
Polyclonal
Unconjugated
PPARGC1A
PPARGC1A, LEM6, PGC1, PGC1A, PGC-1v, PPARGC1, LEM6, Peroxisome proliferator-activated receptor gamma coactivator 1-alpha, Ligand effect modulator 6, PGC-1-alpha
Rabbit
Antigen affinity chromatography
RUO
10891, 19017, 83516
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9UBK2, O70343, Q9QYK2
PPARGC1A
19 amino acid peptide near the carboxy terminus of human PPARGC1A
100 μg
Primary
Human, Mouse, Rat