missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PEX19 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA522129

Product Code. 13219628

  • £504.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: Jurkat, Raji, NCI-H929, K562, THP-1, HL-60, mouse liver, PC-12, Rat-2. Predicted reactivity: Mouse (91%), Rat (93%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is necessary for early peroxisomal biogenesis. It acts both as a cytosolic chaperone and as an import receptor for peroxisomal membrane proteins (PMPs). Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene are a cause Zellweger syndrome (ZWS).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PEX19
Polyclonal
Unconjugated
PEX19
33 kDa housekeeping protein; D1S2223E; HK33; housekeeping gene, 33kD; OK/SW-cl.22; PBD12A; peroxin-19; Peroxisomal biogenesis factor 19; peroxisomal farnesylated protein; peroxisome biogenesis factor 19; Pex19; PMP1; PMPI; PxF; PXMP1
Rabbit
Antigen affinity chromatography
RUO
19298, 289233, 5824
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.01 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
P40855, Q8VCI5, Q9QYU1
PEX19
Recombinant protein encompassing a sequence within the center region of human PEX19. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.