Learn More
Peripherin, clone: 7C5, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen P21935
Description
Peripherin is a 57kD type III intermediate filament that is a specific marker for peripheral neurons, including enteric ganglion cells. Peripherin is expressed in the developing peripheral nervous system and is highly enriched in neuronal derivatives of the neural crest. Peripherin offers an advantage over other neural markers, such as S100 and NSE in that it does not stain chrmaffin cell types.
Specifications
| Peripherin | |
| Monoclonal | |
| Unconjugated | |
| PRPH | |
| NEF4, PRPH1, neurofilament 4 (57kD) | |
| Mouse | |
| IgG fraction | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunofluorescence, Western Blot | |
| 7C5 | |
| Antibody with 0.1% sodium azide | |
| A6QQJ3, P41219, P15331, P21807 | |
| PRPH | |
| Recombinant rat peripherin expressed in and purified from E. coli. | |
| 100 μL | |
| 5630 | |
| Store at 2-8°C for up to 1 month. For long term storage Store at -20°C | |
| Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.