missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Human Perforin, Clone: δG9; PE, BD
Brand: BD Biosciences 563763
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: deltaG9
Isotype: IgG2b, κ
Species Reactivity: Hu
Immunogen: Purified Granules from the Human Lymphoma Cell Line YT
Flow Cytometry (Intracellular Staining)
Specifications
| Perforin | |
| deltaG9 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human |
| Monoclonal | |
| PE-CF594 | |
| Purified Granules from the Human Lymphoma Cell Line YT | |
| 50 Tests | |
| Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction