Learn More
PDYN, Mouse, Clone: 2E12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PDYN.
Brand: Abnova H00005173-M01.100ug
Description
The protein encoded by this gene is a preproprotein that is proteolytically processed to form the secreted opioid peptides beta-neoendorphin, dynorphin, leu-enkephalin, rimorphin, and leumorphin. These peptides are ligands for the kappa-type of opioid receptor. Dynorphin is involved in modulating responses to several psychoactive substances, including cocaine. [provided by RefSeq
Sequence: KRYGGFLRRIRPKLKWDNQKRYGGFLRRQFKVVTRSQEDPNAYSGELFDASpecifications
| PDYN | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_024411 | |
| PDYN | |
| PDYN (NP_077722.1, 205 a.a. ∼ 254 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunoprecipitation, Western Blot | |
| 2E12 | |
| Mouse monoclonal antibody raised against a partial recombinant PDYN. | |
| PDYN | |
| MGC26418/PENKB | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5173 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.