missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDX1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA511897

Product Code. 10785482

  • £477.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Beskrivning

Beskrivning

This antibody is predicted to react with mouse and rat based on sequence homology.

The protein encoded by this gene is a transcriptional activator of several genes, including insulin, somatostatin, glucokinase, islet amyloid polypeptide, and glucose transporter type 2. The encoded nuclear protein is involved in the early development of the pancreas and plays a major role in glucose-dependent regulation of insulin gene expression. Defects in this gene are a cause of pancreatic agenesis, which can lead to early-onset insulin-dependent diabetes mellitus (NIDDM), as well as maturity onset diabetes of the young type 4 (MODY4).
TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

PDX1
Polyclonal
Unconjugated
PDX1
DLDBP; E3BP; Glucose-sensitive factor; GSF; Idx1; IDX-1; Insulin promoter factor 1; insulin promoter factor 1, homeodomain transcription factor; insulin promotor factor 1; insulin upstream factor 1; Ipf1; IPF-1; Islet/duodenum homeobox 1; islet/duodenum homeobox-1; IUF1; IUF-1; MODY4; PAGEN1; Pancreas/duodenum homeobox protein 1; pancreatic and duodenal homeobox 1; pancreatic and duodenal homeobox gene 1; pancreatic-duodenal homeobox factor 1; PDHX; PDX1; pdx-1; somatostatin transactivating factor 1; somatostatin transcription factor 1; somatostatin-transactivating factor 1; Stf1; STF-1
Rabbit
Antigen affinity chromatography, Protein A
RUO
3651
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.25 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P52945
PDX1
KLH conjugated synthetic peptide between 1-30 amino acids from human PDX1.
400 μL
Primary
Human
Antibody
IgG
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Produktspecifikationer
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.