Learn More
anti-PDX1, Polyclonal, Abcam™
Brand: Abcam AB47267-100UL
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Cow, Dog, Human
Immunogen: Recombinant fusion protein containing N-Terminal region of mouse Pdx1
Applications:
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
| PDX1 | |
| Unconjugated | |
| Recombinant fusion protein containing N-Terminal region of mouse Pdx1 | |
| Cancer and Tumor Biology, Developmental Biology, Signal Transduction, Stem Cell Biology | |
| Bovine, Canine, Human, Mouse, Rat | |
| IgG |
| Polyclonal | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Antisera |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.