Learn More
PDPK1 (pS241) Mouse, Unlabeled, Clone: J66-653.44.22, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558395
Description
The serine/threonine kinase 3-Phosphoinositide-Dependent Protein Kinase-1 (PDPK1, also known as PDK1) contributes to the activation of many important kinases in the insulin and IGF-1 signaling pathways. It acts downstream of phosphatidylinositol 3-kinase (PI3-kinase) to phosphorylate residues in the activation loops of many cellular kinases, including protein kinase B (PKB/Akt), PKC isoforms, p70 S6 kinase, and PDPK1 itself. The autophosphorylation of PDPK1 at serine 241 (S241) has recently been suggested to play a role in the regulation of PDPK1. It has been proposed that PDPK1 activity plays a key role in the regulation of various cellular events such as cell proliferation, differentiation, and apoptosis. The J666-653.44.22 monoclonal antibody recognizes the phosphorylated S241 in the activation loop of human PDPK1. The orthologous phosphorylation site in mouse and rat PDPK1 is S244.
Western Blotting
Spécification
| PDPK1 (pS241) | |
| Monoclonal | |
| 0.5mg/mL | |
| PDK1 (pS241), PKB Kinase (pS241) | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Western Blot | |
| J66-653.44.22 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Phosphorylated Human PDPK1 Peptide | |
| 0.1 mg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.