missing translation for 'onlineSavingsMsg'
Learn More

PDI Mouse, Unlabeled, Clone: 34, BD

Mouse Monoclonal Antibody

£219.86 - £284.36

Specifications

Antigen PDI
Clone 34
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15875719
View Documents
BD Biosciences
610946
50 μg
ONLINE EXCLUSIVE PRICE
£229.00
£219.86 / 1 set
Save £9.14
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15885719
View Documents
BD Biosciences
610947
150 μg
ONLINE EXCLUSIVE PRICE
£297.00
£284.36 / 1 set
Save £12.64
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ER is the site of translation of membrane and secretory proteins. Following synthesis, it shuttles these proteins through a contiguous membrane system to their appropriate destinations. Protein disulfide isomerase (PDI) is an abundant, multifunctional, eukaryotic protein. Although it exhibits ubiquitous expression, it is primarily located in the ER lumen. Here, it functions to catalyze the isomerization of intramolecular disulfide bridges, thereby allowing them to generate their most thermodynamically stable configurations. This role in rearrangement has lead to the classification of PDI as a chaperone. Although protein folding occurs in its absence, PDI may be essential for it to proceed at a physiological relevant rate. In addition, PDI is the β-subunit of prolyl 4- hydroxylase and is a component of the triglyceride transfer complex. PDI is retained in the ER lumen via its C-terminal -KdaEL sequence. Via this sequence, it is continuously recycled back to the ER from other membranous compartments. Thus, PDI is a diverse protein whose primary function may be to correct disulfide bonding and, thus, ensure the most stable conformation of newly synthesized proteins.

Immunofluorescence, Western Blotting

Specifications

Specifications

PDI
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
34
Western Blot
Unconjugated
RUO
Canine, Human, Mouse, Rat
Bovine PDI aa. 109-214
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.