missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDE3B Monoclonal Antibody (4A4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA519555
This item is not returnable.
View return policy
Description
Peptide Sequence: LTPFPGFYPC SEIEDPAEKG DRKLNKGLNR NSLPTPQLRR SSGTSGLLPV EQSSRWDRNN GKRPHQEFGI SSQGCYLNGP FNSNLLTIPK QRSSSVSLTH.
Cyclic nucleotide phosphodiesterase with a dual-specificity for the second messengers cAMP and cGMP, which are key regulators of many important physiological processes. May play a role in fat metabolism. Regulates cAMP binding of RAPGEF3. Through simultaneous binding to RAPGEF3 and PIK3R6 assembles a signaling complex in which the PI3K gamma complex is activated by RAPGEF3 and which is involved in angiogenesis.
Specifications
| PDE3B | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q13370 | |
| PDE3B | |
| PDE3B (NP_000913, 401 a.a. ~ 500 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4A4 | |
| Unconjugated | |
| PDE3B | |
| 9830102A01Rik; AI847709; CGIP1; CGI-PDE B; CGIPDE1; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; LOW QUALITY PROTEIN: cGMP-inhibited 3',5'-cyclic phosphodiesterase B; cyclic GMP-inhibited phosphodiesterase B; cyclic nucleotide phosphodiesterase; HcGIP1; LOW QUALITY PROTEIN: cGMP-inhibited 3',5'-cyclic phosphodiesterase B; PDE3B; phosphodiesterase 3B; phosphodiesterase 3B cGMP inhibited; phosphodiesterase 3B, cGMP-inhibited | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5140 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur