missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phosphodiesterase 1B, calmodulin-dependent, Mouse, Clone: 4F9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PDE1B.
Brand: Abnova H00005153-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: TDVAEKSVQPLADEDSKSKNQPSFQWRQPSLDVEVGDPNPDVVSFRSTWVKRIQENKQKWKERAASGITNQMSIDELSPCEEEAPPSPAEDEHNQNGNLDSpecifications
| phosphodiesterase 1B, calmodulin-dependent | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC032226 | |
| PDE1B | |
| PDE1B (AAH32226, 437 a.a. ∼ 536 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Apoptosis | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4F9 | |
| Mouse monoclonal antibody raised against a partial recombinant PDE1B. | |
| PDE1B | |
| PDE1B1/PDES1B | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 5153 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction