missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-PDCD1LG2 Matched Antibody Pair, Abnova™
Mached Pair Antibody
Brand: Abnova H00080380-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-PDCD1LG2 (100μg); Detection antibody: biotinylated mouse monoclonal anti-PDCD1LG2, IgG1 Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Host Species: Mouse, Mouse
Isotype: detection antibody: IgG1, κ
Cross Reactivity: Human
ELISA Pair
Specifications
| PDCD1LG2 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PDCD1LG2. | |
| B7DC/Btdc/CD273/MGC142238/MGC142240/PD-L2/PDCD1L2/PDL2/bA574F11.2 | |
| Mouse | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-PDCD1LG2 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-PDCD1LG2, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Biotin | |
| PDCD1LG2 | |
| PDCD1LG2 | |
| 1 Set | |
| 80380 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction