Learn More
DiscoverX Anti-PD1 Antibody [NAT105]
Anti-PD1 Antibody [NAT105] is an antibody acting as an agonist for PD-1 dimerization and an antagonist for PD-1 signaling. Provided as 100μg of antibody in 100μL buffer.
Brand:  DiscoverX 92-1290
Encompass_Preferred
Product Code. 17302609
  • £964.94 / 100µg
Description
Anti-PD1 Antibody [NAT105] is a mouse monoclonal antibody that detects PD1 (programmed cell death 1) protein (also known as CD279). The antibody inhibits the inhibitory receptor PD1, potentiates antitumor T-cell response in tumor cells, and prevents PD1 anti-apoptotic activity through inhibition of T-cell anti-tumor activity. Provided as 100μg of antibody in 100μL buffer.
Specifications
Specifications
100 μg
Control Ligands